1-Aminopyridinium iodide 98%

BD56482-100G BD56482-500G
BLDPBD56482-100GEA 375 EUR
BLDPBD56482-100G BLDPBD56482-500G
1-Aminopyridinium iodide 98%
1-Aminopyridinium iodide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₇IN₂
MW: 222.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051893
CAS Number: 6295-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR