5-Chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione hydrochloride 98%
|
|
BD566141-5G
BD566141-100MG
BLDPBD566141-5GEA
276
EUR
BLDPBD566141-5G
BLDPBD566141-100MG
5-Chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione hydrochloride 98%
5-Chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₁₂Cl₂N₄O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01571384 CAS Number: 183204-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...