2-(5-Nitropyridin-2-yl)-1,2,3,4-tetrahydroisoquinoline 95%
|
|
BD571771-10G
BD571771-25G
BLDPBD571771-10GEA
1190
EUR
BLDPBD571771-10G
BLDPBD571771-25G
2-(5-Nitropyridin-2-yl)-1,2,3,4-tetrahydroisoquinoline 95%
2-(5-Nitropyridin-2-yl)-1,2,3,4-tetrahydroisoquinoline
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₄H₁₃N₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01309003 CAS Number: 416885-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...