1-Bromo-4-chloro-2-iodobenzene 98%

BD57469-1G BD57469-100G
BLDPBD57469-1GEA 7.4 EUR
BLDPBD57469-1G BLDPBD57469-100G
1-Bromo-4-chloro-2-iodobenzene 98%
1-Bromo-4-chloro-2-iodobenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₃BrCl
Storage Temperature: Refrigerator
MDL Number: MFCD00672943
CAS Number: 148836-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR