1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate 98%
|
|
BD58257-10G
BD58257-25G
BLDPBD58257-10GEA
35.7
EUR
BLDPBD58257-10G
BLDPBD58257-25G
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate 98%
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₇H₁₁F₃N₂O₃S Storage Temperature: Ambient |
MDL Number:
MFCD00210008 CAS Number: 145022-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...