1H,1H,11H-Eicosafluoro-1-undecanol 94%

BD58432-500G BD58432-25G
BLDPBD58432-500GEA 617 EUR
BLDPBD58432-500G BLDPBD58432-25G
1H,1H,11H-Eicosafluoro-1-undecanol 94%
1H,1H,11H-Eicosafluoro-1-undecanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₄F₂₀O
Storage Temperature: Ambient
MDL Number: MFCD00039628
CAS Number: 307-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR