5-(Bromomethyl)benzo[d]thiazole 95%

BD586680-1G BD586680-100MG
BLDPBD586680-1GEA 486 EUR
BLDPBD586680-1G BLDPBD586680-100MG
5-(Bromomethyl)benzo[d]thiazole 95%
5-(Bromomethyl)benzo[d]thiazole

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₈H₆BrNS
Storage Temperature: Freezer
MDL Number: MFCD28659949
CAS Number: 131337-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR