1H-Imidazole-4-carbonitrile 98%

BD58761-10G BD58761-5G
BLDPBD58761-10GEA 128 EUR
BLDPBD58761-10G BLDPBD58761-5G
1H-Imidazole-4-carbonitrile 98%
1H-Imidazole-4-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Formula: C₄H₃N₃
Storage Temperature: Refrigerator
MDL Number: MFCD00233358
CAS Number: 57090-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR