3-Bromo-1-methyl-1H-pyrazole 98%

BD58901-25G BD58901-1G
BLDPBD58901-25GEA 325 EUR
BLDPBD58901-25G BLDPBD58901-1G
3-Bromo-1-methyl-1H-pyrazole 98%
3-Bromo-1-methyl-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C4H5BrN2
Storage Temperature: Ambient
MDL Number: MFCD11110668
CAS Number: 151049-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR