4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine 98%

BD58985-25G BD58985-100G
BLDPBD58985-25GEA 224 EUR
BLDPBD58985-25G BLDPBD58985-100G
4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₆ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD09746275
CAS Number: 71149-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR