2-Methyl-1-(o-tolyl)propan-1-one 95%
|
|
BD593645-100MG
BD593645-250MG
BLDPBD593645-100MGEA
111
EUR
BLDPBD593645-100MG
BLDPBD593645-250MG
2-Methyl-1-(o-tolyl)propan-1-one 95%
2-Methyl-1-(o-tolyl)propan-1-one
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₁H₁₄O Storage Temperature: Ambient |
MDL Number:
MFCD12153366 CAS Number: 2040-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...