1-Methyl-1H-pyrrolo[2,3-b]pyridine 95%

BD59368-25G BD59368-1G
BLDPBD59368-25GEA 853 EUR
BLDPBD59368-25G BLDPBD59368-1G
1-Methyl-1H-pyrrolo[2,3-b]pyridine 95%
1-Methyl-1H-pyrrolo[2,3-b]pyridine

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₈N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09743451
CAS Number: 27257-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR