1-Methylcyclohexanol 97%

BD59585-25G BD59585-10G
BLDPBD59585-25GEA 25.2 EUR
BLDPBD59585-25G BLDPBD59585-10G
1-Methylcyclohexanol 97%
1-Methylcyclohexanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00003857
CAS Number: 590-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR