5-Oxo-5-(3-oxopiperazin-1-yl)pentanoic acid 97%

BD599087-5G BD599087-1G
BLDPBD599087-5GEA 103 EUR
BLDPBD599087-5G BLDPBD599087-1G
5-Oxo-5-(3-oxopiperazin-1-yl)pentanoic acid 97%
5-Oxo-5-(3-oxopiperazin-1-yl)pentanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD09045202
CAS Number: 926237-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR