N-(2-(2-Hydroxy-5-methoxy-1H-indol-3-yl)ethyl)acetamide 95%

BD601150-250MG BD601150-100MG
BLDPBD601150-250MGEA 437 EUR
BLDPBD601150-250MG BLDPBD601150-100MG
N-(2-(2-Hydroxy-5-methoxy-1H-indol-3-yl)ethyl)acetamide 95%
N-(2-(2-Hydroxy-5-methoxy-1H-indol-3-yl)ethyl)acetamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30187118
CAS Number: 229018-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR