2-(1-Hydroxyethyl)pyridine 98%

BD60378-10G BD60378-5G
BLDPBD60378-10GEA 88.2 EUR
BLDPBD60378-10G BLDPBD60378-5G
2-(1-Hydroxyethyl)pyridine 98%
2-(1-Hydroxyethyl)pyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉NO
Storage Temperature: Ambient
MDL Number: MFCD06245419
CAS Number: 18728-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR