Fmoc-3-amino-4-methoxybenzoic acid 97%
|
|
BD604442-250MG
BD604442-5G
BLDPBD604442-250MGEA
16.8
EUR
BLDPBD604442-250MG
BLDPBD604442-5G
Fmoc-3-amino-4-methoxybenzoic acid 97%
Fmoc-3-amino-4-methoxybenzoic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₂₃H₁₉NO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD02682216 CAS Number: 256935-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...