Ethyl-4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate 97%
|
|
BD604704-100MG
BD604704-250MG
BLDPBD604704-100MGEA
91.4
EUR
BLDPBD604704-100MG
BLDPBD604704-250MG
Ethyl-4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate 97%
Ethyl-4-(cyclopropylamino)-2-(methylthio)pyrimidine-5-carboxylate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₁₁H₁₅N₃O₂S Storage Temperature: Ambient |
MDL Number:
MFCD27955205 CAS Number: 651734-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...