3'-Chloro-4'-hydroxyacetophenone 98%
|
|
BD6098-5G
BD6098-10G
BLDPBD6098-5GEA
149
EUR
BLDPBD6098-5G
BLDPBD6098-10G
3'-Chloro-4'-hydroxyacetophenone 98%
3'-Chloro-4'-hydroxyacetophenone
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₇ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00663621 CAS Number: 2892-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...