3-Chloro-4-methoxybenzoic acid 98%
|
|
BD6112-1G
BD6112-10G
BD6112-100G
BLDPBD6112-1GEA
6.3
EUR
BLDPBD6112-1G
BLDPBD6112-10G
BLDPBD6112-100G
3-Chloro-4-methoxybenzoic acid 98%
3-Chloro-4-methoxybenzoic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₇ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00016512 CAS Number: 37908-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...