6-(Methoxycarbonyl)picolinic acid 97%

BD6218-100G BD6218-1G BD6218-5G
BLDPBD6218-100GEA 710 EUR
BLDPBD6218-100G BLDPBD6218-1G BLDPBD6218-5G
6-(Methoxycarbonyl)picolinic acid 97%
6-(Methoxycarbonyl)picolinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C8H7NO4
Storage Temperature: Ambient
MDL Number: MFCD00806109
CAS Number: 7170-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR