3-Bromo-9,9'-spirobi[fluorene] 98%

BD625739-1G BD625739-25G
BLDPBD625739-1GEA 28.4 EUR
BLDPBD625739-1G BLDPBD625739-25G
3-Bromo-9,9'-spirobi[fluorene] 98%
3-Bromo-9,9'-spirobi[fluorene]

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₅H₁₅Br
Storage Temperature: Ambient
MDL Number: MFCD28137969
CAS Number: 1361227-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR