N-[4-(1-Naphthyl)phenyl]-4-biphenylamine 98%

BD627650-1G BD627650-25G
BLDPBD627650-1GEA 11.6 EUR
BLDPBD627650-1G BLDPBD627650-25G
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine 98%
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₈H₂₁N
Storage Temperature: Ambient
MDL Number: MFCD28138088
CAS Number: 897921-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR