Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate 97%
BD628056-5G
BD628056-500G
BLDPBD628056-5GEA
10.5
EUR
BLDPBD628056-5G
BLDPBD628056-500G
Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate 97%
Ethyl 3-(4-(methylamino)-3-nitro-N-(pyridin-2-yl)benzamido)propanoate
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₁₈H₂₀N₄O₅ |
MDL Number:
MFCD09833623 CAS Number: 429659-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...