IC261 98%

BD630486-100MG BD630486-1G
BLDPBD630486-100MGEA 105 EUR
BLDPBD630486-100MG BLDPBD630486-1G
IC261 98%
IC261

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₈H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00118156
CAS Number: 186611-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR