MCC950 sodium salt 98%
|
|
BD630595-10MG
BD630595-1MG
BLDPBD630595-10MGEA
131
EUR
BLDPBD630595-10MG
BLDPBD630595-1MG
MCC950 sodium salt 98%
MCC950 sodium salt
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₂₀H₂₃N₂NaO₅S Storage Temperature: Ambient |
MDL Number:
MFCD30478884 CAS Number: 256373-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...