2-(Diethylamino)ethyl methacrylate 97% stabilised


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

BD63079-500G BD63079-100G
BLDPBD63079-500GEA 42 EUR
BLDPBD63079-500G BLDPBD63079-100G
2-(Diethylamino)ethyl methacrylate 97% stabilised
2-(Diethylamino)ethyl methacrylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00038314
CAS Number: 105-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR