(2-Aminophenyl)acetonitrile 97%

BD6333-10G BD6333-25G
BLDPBD6333-10GEA 208 EUR
BLDPBD6333-10G BLDPBD6333-25G
(2-Aminophenyl)acetonitrile 97%
(2-Aminophenyl)acetonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00091082
CAS Number: 2973-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR