[(R,R)-Teth-TsDPEN-RuCl] 98%

BD634418-100MG BD634418-1G
BLDPBD634418-100MGEA 111 EUR
BLDPBD634418-100MG BLDPBD634418-1G
[(R,R)-Teth-TsDPEN-RuCl] 98%
[(R,R)-Teth-TsDPEN-RuCl]

Storage condition: Inert atmosphere, 2-8°C

Formula: C₃₀H₃₁ClN₂O₂RuS
Storage Temperature: Refrigerator
MDL Number: MFCD16294993
CAS Number: 1192620-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR