2-Amino-4-methoxybenzothiazole 95%
|
|
BD6365-25G
BD6365-100MG
BLDPBD6365-25GEA
181
EUR
BLDPBD6365-25G
BLDPBD6365-100MG
2-Amino-4-methoxybenzothiazole 95%
2-Amino-4-methoxybenzothiazole
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₈H₈N₂OS Storage Temperature: Ambient |
MDL Number:
MFCD00005792 CAS Number: 5464-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...