6,7-Dihydro-4(5H)-benzofuranone 98%

BD6384-5G BD6384-10G
BLDPBD6384-5GEA 67.2 EUR
BLDPBD6384-5G BLDPBD6384-10G
6,7-Dihydro-4(5H)-benzofuranone 98%
6,7-Dihydro-4(5H)-benzofuranone

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₈H₈O₂
Storage Temperature: Freezer
MDL Number: MFCD00215511
CAS Number: 16806-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR