Methyl-3,5-dihydroxybenzoate 98%

BD6422-1000G BD6422-500G
BLDPBD6422-1000GEA 299 EUR
BLDPBD6422-1000G BLDPBD6422-500G
Methyl-3,5-dihydroxybenzoate 98%
Methyl 3,5-dihydroxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00002289
CAS Number: 2150-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR