2,2-Bis(3-amino-4-hydroxylphenyl)propane 98%

BD64284-25G BD64284-10G
BLDPBD64284-25GEA 44.1 EUR
BLDPBD64284-25G BLDPBD64284-10G
2,2-Bis(3-amino-4-hydroxylphenyl)propane 98%
2,2-Bis(3-amino-4-hydroxylphenyl)propane

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₅H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00437413
CAS Number: 1220-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR