2,2-Difluorobenzo[1,3]dioxole-4-boronic acid 98%

BD64440-250MG BD64440-1G
BLDPBD64440-250MGEA 124 EUR
BLDPBD64440-250MG BLDPBD64440-1G
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid 98%
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₅BF₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD08062343
CAS Number: 126120-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR