Methyl-2-cyano-2-methylpropanoate 97%

BD64566-100G BD64566-25G BD64566-10G
BLDPBD64566-100GEA 215 EUR
BLDPBD64566-100G BLDPBD64566-25G BLDPBD64566-10G
Methyl-2-cyano-2-methylpropanoate 97%
Methyl-2-cyano-2-methylpropanoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD03428328
CAS Number: 72291-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR