3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate 95%

BD650351-250MG BD650351-1G
BLDPBD650351-250MGEA 74.6 EUR
BLDPBD650351-250MG BLDPBD650351-1G
3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate 95%
3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₁₅F₃O₄SSi
Storage Temperature: Refrigerator
MDL Number: MFCD10566916
CAS Number: 217813-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR