Methyl-4-aminobenzoate 98%

BD6506-5G BD6506-25G
BLDPBD6506-5GEA 7.4 EUR
BLDPBD6506-5G BLDPBD6506-25G
Methyl-4-aminobenzoate 98%
Methyl 4-aminobenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00007891
CAS Number: 619-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR