Methyl-2-methylnicotinate 97%

BD6522-1G BD6522-25G
BLDPBD6522-1GEA 7.4 EUR
BLDPBD6522-1G BLDPBD6522-25G
Methyl-2-methylnicotinate 97%
Methyl 2-methylnicotinate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00051961
CAS Number: 65719-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR