3-Fluorobenzene-1,2-diamine 98%
|
|
BD65338-5G
BD65338-25G
BLDPBD65338-5GEA
133
EUR
BLDPBD65338-5G
BLDPBD65338-25G
3-Fluorobenzene-1,2-diamine 98%
3-Fluorobenzene-1,2-diamine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C6H7FN2 Storage Temperature: Ambient |
MDL Number:
MFCD00061130 CAS Number: 18645-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...