2-Oxo-2,3-dihydrobenzooxazole-6-sulphonyl chloride 98%

BD65672-5G BD65672-10G
BLDPBD65672-5GEA 237 EUR
BLDPBD65672-5G BLDPBD65672-10G
2-Oxo-2,3-dihydrobenzooxazole-6-sulphonyl chloride 98%
2-Oxo-2,3-dihydrobenzooxazole-6-sulphonyl chloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₄ClNO₄S
Storage Temperature: Ambient
MDL Number: MFCD03001119
CAS Number: 27685-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR