Pinacol 98%

BD65808-500G BD65808-100G
BLDPBD65808-500GEA 57.8 EUR
BLDPBD65808-500G BLDPBD65808-100G
Pinacol 98%
Pinacol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00004462
CAS Number: 76-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR