3-Amino-3-(4-chlorophenyl)propionic acid 97%

BD6590-1G BD6590-10G
BLDPBD6590-1GEA 11.6 EUR
BLDPBD6590-1G BLDPBD6590-10G
3-Amino-3-(4-chlorophenyl)propionic acid 97%
3-Amino-3-(4-chlorophenyl)propionic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126370
CAS Number: 19947-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR