2,4,6-Tribromoresorcinol 98%

BD66203-100G BD66203-1G
BLDPBD66203-100GEA 995 EUR
BLDPBD66203-100G BLDPBD66203-1G
2,4,6-Tribromoresorcinol 98%
2,4,6-Tribromoresorcinol

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₃Br₃O₂
MW: 346.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009717
CAS Number: 2437-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR