2,4-Dichloro-5-(chloromethyl)pyridine 95%

BD66649-250MG BD66649-1G
BLDPBD66649-250MGEA 45.2 EUR
BLDPBD66649-250MG BLDPBD66649-1G
2,4-Dichloro-5-(chloromethyl)pyridine 95%
2,4-Dichloro-5-(chloromethyl)pyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₄Cl₃N
Storage Temperature: Refrigerator
MDL Number: MFCD04114263
CAS Number: 73998-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR