2,6-Diamino-3,5-difluoropyridine 98%
|
|
BD68001-5G
BD68001-1G
BLDPBD68001-5GEA
68.3
EUR
BLDPBD68001-5G
BLDPBD68001-1G
2,6-Diamino-3,5-difluoropyridine 98%
2,6-Diamino-3,5-difluoropyridine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₅H₅F₂N₃ MW: 145.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412208 CAS Number: 247069-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...