2,6-Dichlorodiphenylamine 98%

BD68178-5G BD68178-25G
BLDPBD68178-5GEA 8.4 EUR
BLDPBD68178-5G BLDPBD68178-25G
2,6-Dichlorodiphenylamine 98%
2,6-Dichlorodiphenylamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₂H₉Cl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00269648
CAS Number: 15307-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR