Lithium 3-(2-(4-benzoylpiperazin-1-yl)-2-oxoacetyl)-4-methoxy-7-(3-methyl-1H-1,2,4-triazol-1-yl)pyrrolo[2,3-c]pyridin-1-ide 97%

BD682316-100MG BD682316-250MG
BLDPBD682316-100MGEA 33.6 EUR
BLDPBD682316-100MG BLDPBD682316-250MG
Lithium 3-(2-(4-benzoylpiperazin-1-yl)-2-oxoacetyl)-4-methoxy-7-(3-methyl-1H-1,2,4-triazol-1-yl)pyrrolo[2,3-c]pyridin-1-ide 97%
Lithium 3-(2-(4-benzoylpiperazin-1-yl)-2-oxoacetyl)-4-methoxy-7-(3-methyl-1H-1,2,4-triazol-1-yl)pyrrolo[2,3-c]pyridin-1-ide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₄H₂₂LiN₇O₄
Storage Temperature: Ambient
MDL Number: MFCD30609551
CAS Number: 1449413-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR