Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate 99%

BD683256-5G BD683256-1G
BLDPBD683256-5GEA 692 EUR
BLDPBD683256-5G BLDPBD683256-1G
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate 99%
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₃₀H₂₄F₁₂N₆P₂Ru
MW: 859.562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042502
CAS Number: 60804-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR