(S)-(+)-2-Phenylglycine methyl ester hydrochloride 98%
|
|
BD6838-100G
BD6838-25G
BLDPBD6838-100GEA
58.8
EUR
BLDPBD6838-100G
BLDPBD6838-25G
(S)-(+)-2-Phenylglycine methyl ester hydrochloride 98%
(S)-(+)-2-Phenylglycine methyl ester hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₉H₁₂ClNO₂ MW: 201.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077158 CAS Number: 15028-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...