2,6-Dimethylpyridine-3,5-dicarboxylic acid 97%

BD68423-1G BD68423-25G
BLDPBD68423-1GEA 45.2 EUR
BLDPBD68423-1G BLDPBD68423-25G
2,6-Dimethylpyridine-3,5-dicarboxylic acid 97%
2,6-Dimethylpyridine-3,5-dicarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00023507
CAS Number: 2602-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR